RNA Extraction Stabilizer Reagent, Omega Bio-Tek
Supplier: OMEGA BIO-TEK CA
R0424-02 R0424-01
CA101319-176EA 486.12 CAD
CA101319-176 CA101319-330
RNA Extraction Stabilizer Reagent, Omega Bio-Tek
Buffers Extraction Buffers
The E.Z.N.A.® RNALock Stabilizer Reagent is an innovative reagent which rapidly lyses cells and tissues to inactivate RNases at room temperature. Simply submerge the samples, such as cell cultures, animal tissues, or white blood cells into the RNALock Stabilizer Reagent. The intact cellular RNA will be preserved even at elevated temperature. Samples can be stored in this reagent in excess of 12 months at −20°C.