Chip Measurement System (CMS) Analyzer, Draeger
Supplier: Dräger
6406470 6406270 6406400 6406410 6406450 6406330 6406510 6406160 6406280 6406040 6406580 6406090 6406030 6406060 6406130 6406040 6406200 6406010 6406140 6406210 6406070 6406490 6406180 6406430 6406520 6406080 6406210 6406120 6406140 6406170 6406340 6406260 6406130 6406580 6406490 6406340 6406460 6406020 6406430 6406520 6406190 6406090 6406170 6406450 6406400 6406050 6406100 6406390 6406070 6406110 6406290 6406010 6406060 6406250 6406420 6406320 6406530 6406370 6406560 6406380 6406200 6406440 6406310 6406120 6406150 6406500 6406110 6406560 6406230 6406310 6406380 6406260 6406220 6406420 6406080 6406440 6406230 6406550 6405300 6406150 6406360 6406470 6406020 6406240 6406550 6406360 6406240 6406330 6406190 6406050 6406410 6406180 6406030 6406250 6405300 6406370 6406390 6406320 6406270 6406460 6406290 6406280 6406220 6406160 6406510 6406100 6406500
CA12777-258EA 1026.88 CAD
CA12777-258 47729-684 47729-686 47729-688 34331-678 CA12777-260 12777-250 12777-252 CA12777-264 CA34331-682 CA12777-262 12777-254 CA34331-684 12777-256 CA12777-268 12777-258 CA12777-266 CA34331-680 12777-260 CA12777-270 12777-262 34331-682 12777-264 CA12777-274 34331-680 12777-266 CA12777-272 12777-268 34331-684 CA47729-678 12777-270 12777-272 CA47729-680 12777-274 80080-910 34330-036 34330-038 34330-032 CA47729-686 47729-678 34330-034 CA47729-688 34330-030 47729-680 CA80080-910 13450-868 34330-824 34330-028 CA34330-038 CA34330-036 34330-024 CA34330-034 34330-026 34330-020 CA34330-032 CA34330-030 34330-022 CA13450-868 CA12777-224 CA34330-824 CA12777-228 CA12777-226 34330-018 CA34330-028 CA34330-026 34330-014 CA34330-024 CA34330-022 34330-016 34330-010 CA34330-020 34330-012 CA12777-230 12777-224 CA12777-234 CA12777-232 12777-226 12777-228 CA12777-238 CA12777-236 CA34330-018 34330-008 CA34330-016 CA34330-014 CA34330-012 CA34330-010 12777-230 CA12777-242 CA12777-240 12777-232 CA12777-246 12777-234 CA12777-244 12777-236 12777-238 CA34330-008 CA12777-248 CA34331-678 12777-240 CA12777-252 12777-242 CA12777-250 12777-244 12777-246 CA12777-256 12777-248 CA12777-254
Chip Measurement System (CMS) Analyzer, Draeger
Gas Detection Systems
This easy-to-use, portable system integrates a variety of substance-specific chemical sensing chips with a handheld reader.
  • Portable, easy to use
  • Integrates variety of substance-specific, chemical-sensing chips with handheld reader
Flow sensor guarantees a constant mass flow of the sample through the capillary in the chip so that measurements are not affected by fluctuations in the atmospheric pressure. It takes gas and vapor spot measurements by combining chemistry and electronics to provide accurate and reliable results.
Ordering information: Chips are calibrated for up to two years. Analyzer can be equipped with a remote sampling unit. For details, contact your Avantor sales representative.