VWR® Contour™ Standing Height Corner Base Cabinet
Supplier: VWR International
CFG-2169-12 CFG-2169-A CWG-2169-A
CFG-2169-12EA 3664.23 CAD
CFG-2169-12 CFG-2169-A CWG-2169-A
VWR® Contour™ Standing Height Corner Base Cabinet
Furniture Casework
Heavy-gauge steel full-frame construction.
  • Smooth, easy-to-maintain surfaces
  • Phosphate-coated with a baked, chemical-resistant, powder-coat finish
  • Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
Cabinet has one cupboard and includes sink-support assembly.
Two rear panels 317/8" each, two front panels 211/4" each
Clearance width 151/16"