VWR® Gas Cylinder Wall Mounts and Supports
Supplier: VWR International
82023-826 82023-830 82023-832 82023-834 82023-836 82023-828
82023-826EA 304.48 CAD
82023-826 82023-830 82023-832 82023-834 82023-836 82023-828
VWR® Gas Cylinder Wall Mounts and Supports
Wall Hooks
Cylinder wall brackets secure 10.2 – 30.5 cm (4 – 12") diameter cylinders using 137.2L x 3.8Wcm (54 x 11/2") polypropylene straps that can withstand up to 1200psi.
  • Polypropylene strap
  • Durable brackets and mounts
The single cylinder adjustable bracket permanently mounts to wall to secure one 10.2 – 35.6 cm (4 – 14") diameter cylinder using a polypropylene strap and security chain. The bench mount bracket holds one 10.2 – 35.6 cm (4 – 14") diameter cylinder using dual mounting posts with swivel feet and nonslip pads to clamp to benches or tabletops from 1 – 4.4 cm (3/8 – 13/4") thick. The cylinder floor stand safely supports one 10.2 – 25.4 cm (4 – 10") diameter cylinder.
Ordering information: Fasteners are not included; contact your Avantor sales representative for more information.