Nalgene® Pipette Jars, Thermo Scientific
Supplier: Thermo Fisher Scientific
5242-0040 5242-0050 5242-0060
36322-086CS 1933.92 CAD
36322-086 36322-100 36322-122
Nalgene® Pipette Jars, Thermo Scientific
Pipettes Pipette Washers
Cross-linked high-density polyethylene pipette jars are ideal for soaking and washing pipettes and glassware, and as large general-purpose containers
Caution: Concentrated sulfuric acid will cause jar to fail. When using sulfuric acid dichromate compound, the acid concentration must be 60% or less. See label on jar. Do not mix sulfuric acid dichromate compound in jar. Keep jar on drain board, in sink, or in container large enough to contain contents in case of leakage.
Tough, stiff, seamless jars resist stress-cracking. All sizes have flanged base for added stability. Breakage isMore Product Information