Technologist Choice™ Woven and Non-Woven Gauze Swabs, Bio Nuclear Diagnostics
Supplier: Bio Nuclear Diagnostics
DIS-022G DIS-023
CA53594-154CS 212.12 CAD
CA53594-154 CA53594-164
Technologist Choice™ Woven and Non-Woven Gauze Swabs, Bio Nuclear Diagnostics
First Aid Supplies Wound Dressings
Open-woven gauze swabs are made from 100% cotton for higher absorption and can be used for multiple wound care applications.
  • Made of a rayon/polyester blend and are ideal for dressing, prepping, and packing purposes
  • Nonsterile
Non-woven gauze swabs have finished edges to minimize loose ends, contamination, and linting.