Sulfinert® Sample Loop, 5 µl, Restek
Supplier: Restek
Sulfinert®
22840
RK22840EA 459.04 CAD
RK22840
Sulfinert® Sample Loop, 5 µl, Restek
Chromatography Supplies Chromatography Sample Loops
Ideal for samples containing low concentrations of sulfur or other active compounds.
  • Sample loop sizes from 5 μL to 5 mL
  • Sulfinert®-treated
Sulfinert® treatment eliminates active sites in the loop for better active compound recovery. Sample loop features 1/16" fittings for W-type valves.