Anti-G4S Linker Rabbit Monoclonal Antibody [clone: S13] (PE (Phycoerythrin))

Supplier: CYTOART, INC. MS
212305 212306
MSPP-212305EA 507.69 CAD
MSPP-212305 MSPP-212306
Anti-G4S Linker Rabbit Monoclonal Antibody [clone: S13] (PE (Phycoerythrin))
Antibodies
This antibody reacts with cell membrane-expressed CARs of varying specificity containing a G4S linker within the scFv of the extracellular domain. The poly-Glycine-Serine (G4S) linker is a kind of synthetic peptide linker sequence that is flexible and unstructured. It is frequently used to link the variable heavy (VH) domain and the variable light (VL) domain of single-chain variable fragments (scFvs) and chimeric antigen receptors (CARs) that have an extracellular domain scFv for recognizing target antigens. The linker is made up of a core pentapeptide sequence, Gly-Gly-Gly-Gly-Ser.

  • High sensitivity
  • Low non-specificity
  • Idiotypic
  • Diagnostic material for CAR-T CDx

Reliable CAR-T CDx, been in helping commercialized CAR-T therapeutics. Immediate shipping if in stock, lead time might be up to 30 days if out of stock pending for reproduction.

Type: Primary
Antigen: G4S Linker
Clonality: Monoclonal
Clone: S13
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: NA
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR