Anti-fmc63 scFv Rabbit Monoclonal Antibody [clone: R19M] (PE (Phycoerythrin))

Supplier: CYTOART, INC. MS
200105 200106
MSPP-200105EA 1020 CAD
MSPP-200105 MSPP-200106AB
Anti-fmc63 scFv Rabbit Monoclonal Antibody [clone: R19M] (PE (Phycoerythrin))
Antibodies
The rabbit monoclonal antibody R19M specifically binds to the scFv region of a CD19-specific mouse monoclonal antibody (mAb, clone FMC63). CD19 antigen is a B-cell specific cell surface antigen, which is expressed in all B-cell lineage malignancies and normal B-cells. The scFv region of FMC63 has been used to develop CD19-specific chimeric antigen receptor (CAR) T cells utilized in clinical trials.

  • High sensitivity
  • Low non-specificity
  • Idiotypic
  • Diagnostic material for CAR-T CDx

Reliable CAR-T CDx, been in helping commercialized CAR-T therapeutics. Immediate shipping if in stock, lead time might be up to 30 days if out of stock pending for reproduction.

Type: Primary
Antigen: fmc63 scFv
Clonality: Monoclonal
Clone: R19M
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR