LabClear® Refillable Inline Gas Filters
Supplier: Labclear
RGF-250-200 RGF-250-400 RGF-125-200 RGF-125-400
32574-509EA 999.08 CAD
32574-509 32574-542 32574-586 32574-622
LabClear® Refillable Inline Gas Filters
Filters Gas Filters
Removes water, oil, and other foreign material from inert gases such as nitrogen, helium, argon, and hydrogen.
  • Body Ø: 2".
  • Length: 7¹/₂" (200 ml capacity), 14" (400 ml capacity)
Contains mixture of indicating DRIERITE® desiccant and 13× molecular sieve. Filter body permits refilling with fresh mixture. Made of clear cast acrylic for easy viewing of desiccant color change from blue to pink with moisture absorption. Exhausted filler is easily removed and replaced from premixed refill kit. Tubular inline configuration with nickel-plated brass, threaded, male tube compression fitting on both ends. Used at 125 psi and 100 °F.