GC Tubing, Ohio Valley
Supplier: Ohio Valley
5115 5135 5165 5185
21507-982EA 555.46 CAD
21507-982 21507-984 21507-986 21507-988
GC Tubing, Ohio Valley
Tubing Chromatography Tubing
Stainless steel and copper tubing is manufactured specifically for gas chromatography.
The premium grade Type 304 stainless steel tubing is specially tempered for easy bending. Tubing comes in 15.2m (50') lengths.