VWR® Disposable Serological Pipettes, Glass
Supplier: VWR International
93000-672 93000-674 93000-676 93000-678 93000-680 93000-682 93000-684 93000-686 93000-688 93000-690 93000-694 93000-696 93000-700 93000-704 93000-706 93000-742 93000-692 93000-698 93000-702
93000-672CS 1947.81 CAD
93000-672 93000-674 93000-676 93000-678 93000-680 93000-682 93000-684 93000-686 93000-688 93000-690 93000-694 93000-696 93000-700 93000-704 93000-706 93000-742 93000-692 93000-698 93000-702
VWR® Disposable Serological Pipettes, Glass
Pipettes Serological Pipettes
VWR® Disposable Serological Pipettes are available in sterile or nonsterile formats, with a variety of packaging options to choose from.
  • Uniform pulled tip, calibrated to deliver
  • Borosilicate glass construction
  • Negative graduations increase versatility
  • Available in sterile or nonsterile formats
These pipettes have a uniform pulled tip and are calibrated to deliver to the tip, with two rings near the top denoting that the small amount remaining in the tip after free delivery has ceased must be blown out and added to the main volume delivered. Negative graduations increase the versatility of the pipet. Pipettes are available sterile and plugged or in bulk packs of nonsterile, non-plugged pipettes. Nonsterile, non-plugged pipettes feature a blue scale. Made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class B glass requirements. Individually wrapped, multi-pack, and canister pack pipettes are also designed from ASTM Specification E714.
Packaging: Individually wrapped pipettes are packaged in color-coded paper/plastic wraps and overpacked in convenient dispensing shelf packs for laboratory benches. Multi-pack pipettes come in color-coded bags of 25 pipettes (1 ml and 2 ml sizes) or 10 pipettes (5 ml and 10 ml sizes). Canister pack pipettes are packaged in a reclosable pack of 50 pieces each sealed in a plastic bag. Bulk pack pipettes are sealed in plastic bags and overpacked in shelf packs.