KIMAX® Reusable Serological Pipettes, Glass, Color-Coded, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
37034B-10 37034B-25 37034B-5 37034B-2
89003-526CS 599.67 CAD
89003-526 89003-530 89003-532 89003-528
KIMAX® Reusable Serological Pipettes, Glass, Color-Coded, Kimble Chase
Pipettes Serological Pipettes
Wide-tip pipettes with large tip opening provide faster delivery and are ideal for use with viscous liquids, slurries, and suspensions
Made from KIMAX-51® borosilicate glass for the highest resistance to chemical attack and scratching. Pipettes are calibrated to deliver to the tip. Top ends are tooled to accept cotton plugging. Scale is permanent brown stain fused into the surface without etching. Pipettes are color-coded for ease in sorting and selection. Designed from ASTM specification E1044, Style III requirements.