Disposable Serological Pipettes, Plastic, Kimble Chase
Supplier: DWK Life Sciences
56900-11100 56900-5110 56900-50510 56900-25210 56800-11100 56900-1110 56900-21100 56900-10110
53384-451CS 410.59 CAD
53384-451 14672-918 14672-916 14672-904 14672-914 14672-900 14672-922 14672-920
Disposable Serological Pipettes, Plastic, Kimble Chase
Pipettes Serological Pipettes
These accurate, plastic pipettes are sterile, plugged, and ready for use.
These color coded pipettes feature a black printed scale with reverse and negative graduations to increase the versatility of use. Designed from ASTM Specification E934, and ASTM Specification E1380 for color coding.
Pipettes feature a uniform molded tip and top ends with cotton plugging. They are calibrated to deliver to the tip, with two rings near the top denoting that the small amount remaining after free delivery must be blown out to obtain total rated capacity.
Ordering information: Packaged individually in paper/plastic inner pack of 25 and overpacked in convenient dispensing shelf packs of 100.