9-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-4,7-dioxanonanoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
9-(Fmoc-amino)-4,7-dioxanonanoic acid, Fmoc-NH-PEG2-CH2CH2COOH, Fmoc-9-amino-4,7-dioxanonanoic acid, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10-trioxa-4-azatridecan-13-oic acid, 3-[2-[2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]ethoxy]ethoxy]propionic acid, 3-[2-[2-(Fmoc-amino)ethoxy]ethoxy]propionic acid
F0999-1G
F0999-200MG
TCF0999-1GEA
358.66
CAD
TCF0999-1G
TCF0999-200MG
9-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-4,7-dioxanonanoic acid ≥98.0% (by HPLC, titration analysis)
9-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-4,7-dioxanonanoic acid
Formula:
C₂₂H₂₅NO₆ MW: 399.44 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD08064306 CAS Number: 872679-70-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...