trans,trans-4-(4-Fluorophenyl)-4'-propylbicyclohexyl ≥98.0% (by GC)

Supplier: TCI America

F1102-1G F1102-5G
TCF1102-1GEA 72.68 CAD
TCF1102-1G TCF1102-5G
trans,trans-4-(4-Fluorophenyl)-4'-propylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-(4-Fluorophenyl)-4'-propylbicyclohexyl
Formula: C₂₁H₃₁F
Melting Pt: 71°C
MDL Number: MFCD11053420
CAS Number: 82832-27-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 85.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR