1,3-Bis(4-bromophenyl)-2-propanone ≥99.0% (by GC)

Supplier: TCI America

B4791-1G B4791-200MG
TCB4791-1GEA 153.26 CAD
TCB4791-1G TCB4791-200MG
1,3-Bis(4-bromophenyl)-2-propanone ≥99.0% (by GC)
1,3-Bis(4-bromophenyl)-2-propanone
Formula: C₁₅H₁₂Br₂O
MDL Number: MFCD03843365
CAS Number: 54523-47-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR