6-Methyl-1,3-benzoxazole-2(3H)-thione ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M3063-1G M3063-5G
TCM3063-1GEA 139.04 CAD
TCM3063-1G TCM3063-5G
6-Methyl-1,3-benzoxazole-2(3H)-thione ≥98.0% (by GC, titration analysis)
6-Methyl-1,3-benzoxazole-2(3H)-thione
Formula: C₈H₇NOS
MDL Number: MFCD00175644
CAS Number: 23417-29-0

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 209.0 to 213.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR