Moxifloxacin hydrochloride monohydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

M2479-100MG M2479-1G
TCM2479-100MGEA 178.43 CAD
TCM2479-100MG TCM2479-1G
Moxifloxacin hydrochloride monohydrate ≥98.0% (by HPLC, titration analysis)
Moxifloxacin hydrochloride monohydrate
Formula: C₂₁H₂₄FN₃O₄·HCl·H₂O
MDL Number: MFCD09836237
CAS Number: 192927-63-2
Merck Index: 14,06291

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -123.5 to -136.5 deg (C=0.5, water)
Water 3.0 to 5.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR