2,2'-Bipyridine-6,6'-diol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B3966-1G B3966-5G
TCB3966-1GEA 210.14 CAD
TCB3966-1G TCB3966-5G
2,2'-Bipyridine-6,6'-diol ≥98.0% (by GC, titration analysis)
2,2'-Bipyridine-6,6'-diol
Formula: C₁₀H₈N₂O₂
Melting Pt: 340°C
MDL Number: MFCD13194839
CAS Number: 103505-54-0

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR