1-(tert-Butoxycarbonyl)guanidine ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 1-Boc-guanidine, Boc-Guanidine

B5001-25G B5001-5G
TCB5001-25GEA 266.07 CAD
TCB5001-25G TCB5001-5G
1-(tert-Butoxycarbonyl)guanidine ≥98.0% (by titrimetric analysis)
1-(tert-Butoxycarbonyl)guanidine
Formula: C₆H₁₃N₃O₂
MW: 159.189 g/mol
Melting Pt: 168°C
MDL Number: MFCD07369706
CAS Number: 219511-71-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR