3-Amino-5-(4-methoxyphenyl)pyrazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

A2953-1G A2953-5G
TCA2953-1GEA 225.94 CAD
TCA2953-1G TCA2953-5G
3-Amino-5-(4-methoxyphenyl)pyrazole ≥98.0% (by HPLC, titration analysis)
3-Amino-5-(4-methoxyphenyl)pyrazole
Formula: C₁₀H₁₁N₃O
MDL Number: MFCD03093928
CAS Number: 19541-95-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 141.0 to 145.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR