1-(2-Chlorophenyl)-3-methyl-5-pyrazolone 98,0 HPLC, T

Supplier: TCI America
Warning

C3414-25G C3414-5G
76218-936EA 145.36 CAD
76218-936 76218-938
1-(2-Chlorophenyl)-3-methyl-5-pyrazolone 98,0 HPLC, T
1-(2-Chlorophenyl)-3-methyl-5-pyrazolone
Formula: C₁₀H₉ClN₂O
MW: 208.65 g/mol
MDL Number: MFCD00059717
CAS Number: 14580-22-4
EINECS: 238-623-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 198.0 to 202.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR