5-Bromo-6-hydroxynicotinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B5193-1G B5193-5G
TCB5193-1GEA 91.64 CAD
TCB5193-1G TCB5193-5G
5-Bromo-6-hydroxynicotinic acid ≥98.0% (by GC, titration analysis)
5-Bromo-6-hydroxynicotinic acid
Formula: C₆H₄BrNO₃
Melting Pt: 296°C
MDL Number: MFCD08235173
CAS Number: 41668-13-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR