4-Hydrazinobenzenesulfonic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

H1662-100G H1662-25G H1662-500G
TCH1662-100GEA 152.68 CAD
TCH1662-100G TCH1662-25G TCH1662-500G
4-Hydrazinobenzenesulfonic acid ≥98.0% (by HPLC, titration analysis)
4-Hydrazinobenzenesulfonic acid
Formula: C₆H₈N₂O₃S
Melting Pt: 286°C
MDL Number: MFCD00025097
CAS Number: 98-71-5
EINECS: 202-694-2
Merck Index: 14,04773

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Drying loss max. 2.0 %
Suitability for p-cresol-analysis to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR