Amodiaquine dihydrochloride dihydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A3133-25G
A3133-5G
TCA3133-25GEA
140.62
CAD
TCA3133-25G
TCA3133-5G
Amodiaquine dihydrochloride dihydrate ≥98.0% (by HPLC, titration analysis)
Amodiaquine dihydrochloride dihydrate
Formula:
C₂₀H₂₂ClN₃O·2HCl·2H₂O |
MDL Number:
MFCD00078857 CAS Number: 6398-98-7 EINECS: 200-706-0 Merck Index: 14,00572 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Water | 7.0 to 9.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...