trans,trans-4-n-Butyl-4'-(3,4-difluorophenyl)bicyclohexyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: trans,trans-4'-Butyl-4-(3,4-difluorophenyl)bicyclohexyl

B4925-25G B4925-5G
TCB4925-25GEA 885.91 CAD
TCB4925-25G TCB4925-5G
trans,trans-4-n-Butyl-4'-(3,4-difluorophenyl)bicyclohexyl ≥98.0% (by GC)
trans,trans-4-n-Butyl-4'-(3,4-difluorophenyl)bicyclohexyl
Formula: C₂₂H₃₂F₂
MW: 334.495 g/mol
MDL Number: MFCD13188634
CAS Number: 82832-58-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 49.0 deg-C
Mesomorphic range 44.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR