Boc-L-norvaline ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B4320-1G B4320-5G
TCB4320-1GEA 33.18 CAD
TCB4320-1G TCB4320-5G
Boc-L-norvaline ≥98.0% (by titrimetric analysis)
Boc-L-norvaline
Formula: C₁₀H₁₉NO₄
MDL Number: MFCD00037268
CAS Number: 53308-95-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 64.0 to 69.0 deg-C
Specific rotation [a]20/D -14.0 to -17.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR