Boc-Abu-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
B5115-25G
B5115-5G
76218-830EA
162.74
CAD
76218-830
76218-832
Boc-Abu-OH ≥98.0% (by HPLC, titration analysis)
Boc-Abu-OH
Formula:
C₉H₁₇NO₄ |
MDL Number:
MFCD00037267 CAS Number: 34306-42-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 73.0 to 77.0 deg-C |
Specific rotation [a]20/D | -17.0 to -20.0 deg (C=1,MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...