Ethyl 3-Methylpyrazole-5-carboxylate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
3-Methylpyrazole-5-carboxylic acid ethyl ester, Ethyl 3-methyl-1H-pyrazole-5-carboxylate
E1100-1G
E1100-5G
TCE1100-1GEA
33.18
CAD
TCE1100-1G
TCE1100-5G
Ethyl 3-Methylpyrazole-5-carboxylate ≥98.0% (by GC)
Ethyl 3-Methylpyrazole-5-carboxylate
Formula:
C₇H₁₀N₂O₂ MW: 154.165 g/mol |
MDL Number:
MFCD00052514 CAS Number: 1226862-09-4 EINECS: 223-702-0 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...