Thieno[3,2-d]pyrimidin-4(1H)-one ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3H,4H-thieno[3,2-d]pyrimidin-4-one

T3301-1G T3301-5G
TCT3301-1GEA 79 CAD
TCT3301-1G TCT3301-5G
Thieno[3,2-d]pyrimidin-4(1H)-one ≥98.0% (by HPLC, titration analysis)
Thieno[3,2-d]pyrimidin-4(1H)-one
Formula: C₆H₄N₂OS
MW: 152.17 g/mol
MDL Number: MFCD00122140
CAS Number: 16234-10-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 221.0 to 225.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR