(R)-(-)-1-Benzyl-3-hydroxypiperidine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B4857-1G
B4857-5G
TCB4857-1GEA
102.7
CAD
TCB4857-1G
TCB4857-5G
(R)-(-)-1-Benzyl-3-hydroxypiperidine ≥98.0% (by GC)
(R)-(-)-1-Benzyl-3-hydroxypiperidine
Formula:
C₁₂H₁₇NO Boiling Pt: 102°C |
MDL Number:
MFCD00274307 CAS Number: 91599-81-4 |
Specification Test Results
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Specific rotation [a]20/D | -10.0 to -13.0 deg (C=1, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...