1H-Pyrazole-3-carbaldehyde ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
P2261-1G
P2261-200MG
TCP2261-1GEA
461.36
CAD
TCP2261-1G
TCP2261-200MG
1H-Pyrazole-3-carbaldehyde ≥98.0% (by GC)
1H-Pyrazole-3-carbaldehyde
Formula:
C₄H₄N₂O |
MDL Number:
MFCD00129925 CAS Number: 3920-50-1 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 149.0 to 153.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...