1-Methyl-3-octylimidazolium bis(trifluoromethylsulfonyl)imide ≥97.0% (by HPLC)
Supplier: TCI America
|
Warning
|
M3039-25G
M3039-5G
76220-338EA
475.58
CAD
76220-338
76220-340
1-Methyl-3-octylimidazolium bis(trifluoromethylsulfonyl)imide ≥97.0% (by HPLC)
1-Methyl-3-octylimidazolium bis(trifluoromethylsulphonyl)imide
Formula:
C₁₄H₂₃F₆N₃O₄S₂ |
MDL Number:
MFCD08458946 CAS Number: 178631-04-4 |
Specification Test Results
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (HPLC) | min. 97.0 area% |
Purity (Ion exchange titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...