Methyl indol-3-ylacetate ≥98.0% (by GC)

Supplier: TCI America

M2605-25G M2605-5G
TCM2605-25GEA 609.88 CAD
TCM2605-25G TCM2605-5G
Methyl indol-3-ylacetate ≥98.0% (by GC)
Methyl indol-3-ylacetate
Formula: C₁₁H₁₁NO₂
Boiling Pt: 160°C
Storage Temperature: Refrigerator
MDL Number: MFCD00022749
CAS Number: 1912-33-0
EINECS: 217-622-5

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to lump
Purity (GC) min. 98.0 %
Melting point 49.0 to 53.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR