N,N'-Dibutylquinacridone ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

D4780-1G D4780-200MG
TCD4780-1GEA 651.28 CAD
TCD4780-1G TCD4780-200MG
N,N'-Dibutylquinacridone ≥98.0% (by HPLC, total nitrogen)
N,N'-Dibutylquinacridone
Formula: C₂₈H₂₈N₂O₂
Melting Pt: 265°C
MDL Number: MFCD12407159
CAS Number: 99762-80-8

Order Now

Specification Test Results

Appearance Orange to Red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Lambda max. 518.0 to 523.0 nm (CH2Cl2)
Molar extinction coefficient min. 17000 (CH2Cl2, 518.0 to 523.0nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR