4-Phenyldibenzothiophene ≥98.0% (by GC)

Supplier: TCI America

P2510-1G P2510-5G
TCP2510-1GEA 442.4 CAD
TCP2510-1G TCP2510-5G
4-Phenyldibenzothiophene ≥98.0% (by GC)
4-Phenyldibenzothiophene
Formula: C₁₈H₁₂S
MDL Number: MFCD02683728
CAS Number: 98251-31-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR