2-Triphenylenecarboxaldehyde ≥98.0% (by GC)

Supplier: TCI America

T3304-1G T3304-200MG
TCT3304-1GEA 527.72 CAD
TCT3304-1G TCT3304-200MG
2-Triphenylenecarboxaldehyde ≥98.0% (by GC)
2-Triphenylenecarboxaldehyde
Formula: C₁₉H₁₂O
Melting Pt: 162°C
MDL Number: MFCD29089374
CAS Number: 96404-79-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR