N-(tert-Butoxycarbonyl)-D-threonine methyl ester ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
Boc-D-Thr-OMe, N-Boc-D-threonine methyl ester, (2R,3S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-hydroxybutanoate
B5537-1G
B5537-5G
TCB5537-1GEA
151.68
CAD
TCB5537-1G
TCB5537-5G
N-(tert-Butoxycarbonyl)-D-threonine methyl ester ≥98.0% (by HPLC, total nitrogen)
N-(tert-Butoxycarbonyl)-D-threonine methyl ester
Formula:
C₁₀H₁₉NO₅ MW: 233.26 g/mol |
MDL Number:
MFCD01073763 CAS Number: 96099-84-2 |
Specification Test Results
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Specific rotation [a]20/D | +18.0 to +22.0 deg (C=0.5, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...