1-(3-Butenyloxy)-2,3,3,4,4,5,5-heptafluorocyclopentene ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: BHFC

B4881-1G B4881-5G
76219-440EA 507.18 CAD
76219-440 76219-442
1-(3-Butenyloxy)-2,3,3,4,4,5,5-heptafluorocyclopentene ≥97.0% (by GC)
1-(3-Butenyloxy)-2,3,3,4,4,5,5-heptafluorocyclopentene
Formula: C₉H₇F₇O
MW: 264.14 g/mol
MDL Number: MFCD00084878
CAS Number: 958032-89-8

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR