2'-Fluoro-4-pentyl-4''-propyl-1,1':4',1''-terphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2'-Fluoro-4-pentyl-4''-propyl-p-terphenyl

F1046-1G F1046-5G
TCF1046-1GEA 132.25 CAD
TCF1046-1G TCF1046-5G
2'-Fluoro-4-pentyl-4''-propyl-1,1':4',1''-terphenyl ≥98.0% (by GC)
2'-Fluoro-4-pentyl-4''-propyl-1,1':4',1''-terphenyl
Formula: C₂₆H₂₉F
MW: 360.52 g/mol
MDL Number: MFCD11045061
CAS Number: 95759-51-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 51.0 to 55.0 deg-C
Mesomorphic range 50.0 to 143.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR