4-(4-Bromophenyl)dibenzofuran ≥98.0% (by GC)

Supplier: TCI America

B4884-1G B4884-5G
TCB4884-1GEA 123.24 CAD
TCB4884-1G TCB4884-5G
4-(4-Bromophenyl)dibenzofuran ≥98.0% (by GC)
4-(4-Bromophenyl)dibenzofuran
Formula: C₁₈H₁₁BrO
MDL Number: MFCD19441306
CAS Number: 955959-84-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR