1-Phenyl-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-benzimidazole ≥98.0% (by GC, titration analysis)
Supplier: TCI America
P2263-1G
P2263-5G
TCP2263-1GEA
184.86
CAD
TCP2263-1G
TCP2263-5G
1-Phenyl-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-benzimidazole ≥98.0% (by GC, titration analysis)
1-Phenyl-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-benzimidazole
Formula:
C₂₅H₂₅BN₂O₂ |
MDL Number:
MFCD16038225 CAS Number: 952514-86-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 172.0 to 176.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...