(R)-(+)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D3867-5G D3867-1G
76219-026EA 1676.38 CAD
76219-026 76219-696
(R)-(+)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether ≥98.0% (by GC, titration analysis)
(R)-(+)-alpha,alpha-Diphenyl-2-pyrrolidinemethanol trimethylsilyl ether
Formula: C₂₀H₂₇NOSi
Storage Temperature: Refrigerator
MDL Number: MFCD09265100
CAS Number: 943757-71-9

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +53.0 to +57.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR