3-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-[3-(pyridin-3-yl)phenyl]-1,3,2-dioxaborolane, 3-(3-Pyridyl)phenylboronic acid Pinacol ester

T3313-1G T3313-5G
TCT3313-1GEA 270.18 CAD
TCT3313-1G TCT3313-5G
3-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine ≥98.0% (by GC, titration analysis)
3-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine
Formula: C₁₇H₂₀BNO₂
MW: 281.16 g/mol
MDL Number: MFCD20923943
CAS Number: 939430-30-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR