Phenyl(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate ≥93.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: Phenyl(2,4,6-trimethoxyphenyl)iodonium Tosylate

P2412-1G P2412-200MG
TCP2412-1GEA 425.02 CAD
TCP2412-1G TCP2412-200MG
Phenyl(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate ≥93.0% (by HPLC)
Phenyl(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate
Formula: C₂₂H₂₃IO₆S
MW: 542.38 g/mol
Storage Temperature: Refrigerator
CAS Number: 936326-60-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 93.0 % (total of Ion-Pair)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR