3-Bromo-9-(1-naphthyl)-9H-carbazole ≥97.0% (by GC)

Supplier: TCI America
Warning

B5050-1G B5050-200MG
TCB5050-1GEA 102.7 CAD
TCB5050-1G TCB5050-200MG
3-Bromo-9-(1-naphthyl)-9H-carbazole ≥97.0% (by GC)
3-Bromo-9-(1-naphthyl)-9H-carbazole
Formula: C₂₂H₁₄BrN
MDL Number: MFCD26127423
CAS Number: 934545-83-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR