9,10-Dihydroacridine ≥98.0% (by GC)

Supplier: TCI America
Warning

D5058-1G D5058-5G
TCD5058-1GEA 91.64 CAD
TCD5058-1G TCD5058-5G
9,10-Dihydroacridine ≥98.0% (by GC)
9,10-Dihydroacridine
Formula: C₁₃H₁₁N
Boiling Pt: 348°C
MDL Number: MFCD00022256
CAS Number: 92-81-9
EINECS: 202-192-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 168.0 to 172.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR