2,6-Dibromo-N,N'-di-n-octyl-1,8:4,5-naphthalenetetracarboxdiimide ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4,9-Dibromo-2,7-dioctylbenzo[lmn][3,8]phenanthroline-1,3,6,8(2H,7H)-tetraone

D4946-1G D4946-5G
76219-074EA 944.84 CAD
76219-074 76219-076
2,6-Dibromo-N,N'-di-n-octyl-1,8:4,5-naphthalenetetracarboxdiimide ≥98.0% (by HPLC)
2,6-Dibromo-N,N'-di-n-octyl-1,8:4,5-naphthalenetetracarboxdiimide
Formula: C₃₀H₃₆Br₂N₂O₄
MDL Number: MFCD29472527
CAS Number: 926643-78-9

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR