4-(trans-4-Butylcyclohexyl)cyclohexanone ≥98.0% (by GC)
Supplier: TCI America
B4946-25G
B4946-5G
TCB4946-25GEA
181.86
CAD
TCB4946-25G
TCB4946-5G
4-(trans-4-Butylcyclohexyl)cyclohexanone ≥98.0% (by GC)
4-(trans-4-Butylcyclohexyl)cyclohexanone
Formula:
C₁₆H₂₈O |
MDL Number:
MFCD06797514 CAS Number: 92413-47-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...